EOMES Monoclonal Antibody (WD1928), PerCP-eFluor™ 710, eBioscience™ - FAQs
EOMES Monoclonal Antibody (WD1928), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for EOMES Monoclonal Antibody (WD1928), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-4877-41, 46-4877-42)