Syrian Hamster IgG Isotype Control, PerCP-eFluor™ 710, eBioscience™ - FAQs
Syrian Hamster IgG Isotype Control, PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Syrian Hamster IgG Isotype Control, PerCP-eFluor™ 710, eBioscience™ - FAQs (46-4914-80)