CD5 Monoclonal Antibody (YKIX322.3), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD5 Monoclonal Antibody (YKIX322.3), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD5 Monoclonal Antibody (YKIX322.3), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5050-42, 46-5050-41)