BrdU Monoclonal Antibody (BU20A), PerCP-eFluor™ 710, eBioscience™ - FAQs
BrdU Monoclonal Antibody (BU20A), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for BrdU Monoclonal Antibody (BU20A), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5071-41, 46-5071-42)