CD8a Monoclonal Antibody (YCATE55.9), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD8a Monoclonal Antibody (YCATE55.9), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (YCATE55.9), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5080-42, 46-5080-41)