CD85d (ILT4) Monoclonal Antibody (42D1), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD85d (ILT4) Monoclonal Antibody (42D1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD85d (ILT4) Monoclonal Antibody (42D1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5149-41)