CD200 Monoclonal Antibody (OX90), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD200 Monoclonal Antibody (OX90), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD200 Monoclonal Antibody (OX90), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5200-82, 46-5200-80)