CD200 Receptor Monoclonal Antibody (OX110), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD200 Receptor Monoclonal Antibody (OX110), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD200 Receptor Monoclonal Antibody (OX110), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5201-82)