TSLP Receptor Monoclonal Antibody (eBio1A6 (1A6)), PerCP-eFluor™ 710, eBioscience™ - FAQs
TSLP Receptor Monoclonal Antibody (eBio1A6 (1A6)), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for TSLP Receptor Monoclonal Antibody (eBio1A6 (1A6)), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5499-42, 46-5499-41)