Ki-67 Monoclonal Antibody (SolA15), PerCP-eFluor™ 710, eBioscience™ - FAQs
Ki-67 Monoclonal Antibody (SolA15), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Ki-67 Monoclonal Antibody (SolA15), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5698-82, 46-5698-80)