Ki-67 Monoclonal Antibody (20Raj1), PerCP-eFluor™ 710, eBioscience™ - FAQs
Ki-67 Monoclonal Antibody (20Raj1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Ki-67 Monoclonal Antibody (20Raj1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5699-42, 46-5699-41)