MerTK Monoclonal Antibody (DS5MMER), PerCP-eFluor™ 710, eBioscience™ - FAQs
MerTK Monoclonal Antibody (DS5MMER), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for MerTK Monoclonal Antibody (DS5MMER), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5751-80, 46-5751-82)