Mouse IgM Monoclonal Antibody (II/41), PerCP-eFluor™ 710, eBioscience™ - FAQs
Mouse IgM Monoclonal Antibody (II/41), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Mouse IgM Monoclonal Antibody (II/41), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5790-80, 46-5790-82)