TCR V beta 6 Monoclonal Antibody (RR4-7), PerCP-eFluor™ 710, eBioscience™ - FAQs
TCR V beta 6 Monoclonal Antibody (RR4-7), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for TCR V beta 6 Monoclonal Antibody (RR4-7), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5795-82)