TCR V beta 13 Monoclonal Antibody (MR12-3), PerCP-eFluor™ 710, eBioscience™ - FAQs
TCR V beta 13 Monoclonal Antibody (MR12-3), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for TCR V beta 13 Monoclonal Antibody (MR12-3), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5797-80)