TCR V beta 11 Monoclonal Antibody (RR3-15), PerCP-eFluor™ 710, eBioscience™ - FAQs
TCR V beta 11 Monoclonal Antibody (RR3-15), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for TCR V beta 11 Monoclonal Antibody (RR3-15), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5827-80)