TCR V gamma 2 Monoclonal Antibody (UC3-10A6), PerCP-eFluor™ 710, eBioscience™ - FAQs
TCR V gamma 2 Monoclonal Antibody (UC3-10A6), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for TCR V gamma 2 Monoclonal Antibody (UC3-10A6), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5828-82, 46-5828-80)