Siglec-G Monoclonal Antibody (SH2.1), PerCP-eFluor™ 710, eBioscience™ - FAQs
Siglec-G Monoclonal Antibody (SH2.1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Siglec-G Monoclonal Antibody (SH2.1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5833-82, 46-5833-80)