Ly-49E/F Monoclonal Antibody (CM4), PerCP-eFluor™ 710, eBioscience™ - FAQs
Ly-49E/F Monoclonal Antibody (CM4), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Ly-49E/F Monoclonal Antibody (CM4), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5848-82)