CD314 (NKG2D) Monoclonal Antibody (1D11), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD314 (NKG2D) Monoclonal Antibody (1D11), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD314 (NKG2D) Monoclonal Antibody (1D11), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5878-41, 46-5878-42)