NKG2A/C/E Monoclonal Antibody (20d5), PerCP-eFluor™ 710, eBioscience™ - FAQs
NKG2A/C/E Monoclonal Antibody (20d5), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for NKG2A/C/E Monoclonal Antibody (20d5), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5896-82)