NKG2AB6 Monoclonal Antibody (16a11), PerCP-eFluor™ 710, eBioscience™ - FAQs
NKG2AB6 Monoclonal Antibody (16a11), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for NKG2AB6 Monoclonal Antibody (16a11), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5897-80)