FceR1 alpha Monoclonal Antibody (AER-37 (CRA1)), PerCP-eFluor™ 710, eBioscience™ - FAQs
FceR1 alpha Monoclonal Antibody (AER-37 (CRA1)), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for FceR1 alpha Monoclonal Antibody (AER-37 (CRA1)), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5899-41, 46-5899-42)