iNOS Monoclonal Antibody (CXNFT), PerCP-eFluor™ 710, eBioscience™ - FAQs
iNOS Monoclonal Antibody (CXNFT), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for iNOS Monoclonal Antibody (CXNFT), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5920-80, 46-5920-82)