B7-H4 Monoclonal Antibody (H74), PerCP-eFluor™ 710, eBioscience™ - FAQs
B7-H4 Monoclonal Antibody (H74), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for B7-H4 Monoclonal Antibody (H74), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5949-42, 46-5949-41)