Ly-6D Monoclonal Antibody (49-H4), PerCP-eFluor™ 710, eBioscience™ - FAQs
Ly-6D Monoclonal Antibody (49-H4), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Ly-6D Monoclonal Antibody (49-H4), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5974-80, 46-5974-82)