Mouse IgD Monoclonal Antibody (11-26c (11-26)), PerCP-eFluor™ 710, eBioscience™ - FAQs
Mouse IgD Monoclonal Antibody (11-26c (11-26)), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Mouse IgD Monoclonal Antibody (11-26c (11-26)), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5993-80, 46-5993-82)