MHC Class I (H-2Kd/H-2Dd) Monoclonal Antibody (34-1-2S), PerCP-eFluor™ 710, eBioscience™ - FAQs
MHC Class I (H-2Kd/H-2Dd) Monoclonal Antibody (34-1-2S), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for MHC Class I (H-2Kd/H-2Dd) Monoclonal Antibody (34-1-2S), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5998-82, 46-5998-80)