GFP Monoclonal Antibody (5F12.4), PerCP-eFluor™ 710, eBioscience™ - FAQs
GFP Monoclonal Antibody (5F12.4), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for GFP Monoclonal Antibody (5F12.4), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-6498-80)