IkB zeta Monoclonal Antibody (LK2NAP), PerCP-eFluor™ 710, eBioscience™ - FAQs
IkB zeta Monoclonal Antibody (LK2NAP), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IkB zeta Monoclonal Antibody (LK2NAP), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-6801-82, 46-6801-80)