ROR gamma (t) Monoclonal Antibody (B2D), PerCP-eFluor™ 710, eBioscience™ - FAQs
ROR gamma (t) Monoclonal Antibody (B2D), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for ROR gamma (t) Monoclonal Antibody (B2D), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-6981-82, 46-6981-80)