IL-4 Monoclonal Antibody (11B11), PerCP-eFluor™ 710, eBioscience™ - FAQs
IL-4 Monoclonal Antibody (11B11), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IL-4 Monoclonal Antibody (11B11), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-7041-82, 46-7041-80)