IL-6 Monoclonal Antibody (MP5-20F3), PerCP-eFluor™ 710, eBioscience™ - FAQs
IL-6 Monoclonal Antibody (MP5-20F3), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IL-6 Monoclonal Antibody (MP5-20F3), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-7061-82, 46-7061-80)