IL-9 Monoclonal Antibody (MH9D1), PerCP-eFluor™ 710, eBioscience™ - FAQs
IL-9 Monoclonal Antibody (MH9D1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IL-9 Monoclonal Antibody (MH9D1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-7098-42, 46-7098-41)