IL-13 Monoclonal Antibody (eBio13A), PerCP-eFluor™ 710, eBioscience™ - FAQs
IL-13 Monoclonal Antibody (eBio13A), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IL-13 Monoclonal Antibody (eBio13A), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-7133-82, 46-7133-80)