IL-17F Monoclonal Antibody (SHLR17), PerCP-eFluor™ 710, eBioscience™ - FAQs
IL-17F Monoclonal Antibody (SHLR17), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IL-17F Monoclonal Antibody (SHLR17), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-7169-42, 46-7169-41)