IL-22 Monoclonal Antibody (1H8PWSR), PerCP-eFluor™ 710, eBioscience™ - FAQs
IL-22 Monoclonal Antibody (1H8PWSR), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IL-22 Monoclonal Antibody (1H8PWSR), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-7221-82, 46-7221-80)