IL-22 Monoclonal Antibody (22URTI), PerCP-eFluor™ 710, eBioscience™ - FAQs
IL-22 Monoclonal Antibody (22URTI), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IL-22 Monoclonal Antibody (22URTI), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-7229-41)