EBI3 (IL-27 subunit) Monoclonal Antibody (ebic6), PerCP-eFluor™ 710, eBioscience™ - FAQs
EBI3 (IL-27 subunit) Monoclonal Antibody (ebic6), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for EBI3 (IL-27 subunit) Monoclonal Antibody (ebic6), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-7358-42)