IL-17F Monoclonal Antibody (eBio18F10), PerCP-eFluor™ 710, eBioscience™ - FAQs
IL-17F Monoclonal Antibody (eBio18F10), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IL-17F Monoclonal Antibody (eBio18F10), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-7471-80)