IL-23 p19 Monoclonal Antibody (23dcdp), PerCP-eFluor™ 710, eBioscience™ - FAQs
IL-23 p19 Monoclonal Antibody (23dcdp), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IL-23 p19 Monoclonal Antibody (23dcdp), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-7823-41, 46-7823-42)