TL1A Monoclonal Antibody (Tandys1a), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for TL1A Monoclonal Antibody (Tandys1a), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-7911-82, 46-7911-80)