CD217 (IL-17Ra) Monoclonal Antibody (424LTS), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD217 (IL-17Ra) Monoclonal Antibody (424LTS), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD217 (IL-17Ra) Monoclonal Antibody (424LTS), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-7917-42)