IL-8 (1-77) (CXCL8) Monoclonal Antibody (8CH), PerCP-eFluor™ 710, eBioscience™ - FAQs
IL-8 (1-77) (CXCL8) Monoclonal Antibody (8CH), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IL-8 (1-77) (CXCL8) Monoclonal Antibody (8CH), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-8088-41, 46-8088-42)