SSEA4 Monoclonal Antibody (eBioMC-813-70 (MC-813-70)), PerCP-eFluor™ 710, eBioscience™ - FAQs
SSEA4 Monoclonal Antibody (eBioMC-813-70 (MC-813-70)), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for SSEA4 Monoclonal Antibody (eBioMC-813-70 (MC-813-70)), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-8843-41, 46-8843-42)