Granzyme K Monoclonal Antibody (G3H69), PerCP-eFluor™ 710, eBioscience™ - FAQs
Granzyme K Monoclonal Antibody (G3H69), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Granzyme K Monoclonal Antibody (G3H69), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-8897-41, 46-8897-42)