Granzyme B Monoclonal Antibody (NGZB), PerCP-eFluor™ 710, eBioscience™ - FAQs
Granzyme B Monoclonal Antibody (NGZB), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Granzyme B Monoclonal Antibody (NGZB), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-8898-80, 46-8898-82)