CD200 Receptor Monoclonal Antibody (OX108), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD200 Receptor Monoclonal Antibody (OX108), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD200 Receptor Monoclonal Antibody (OX108), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9201-41, 46-9201-42)