Podoplanin Monoclonal Antibody (NZ-1.3), PerCP-eFluor™ 710, eBioscience™ - FAQs
Podoplanin Monoclonal Antibody (NZ-1.3), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Podoplanin Monoclonal Antibody (NZ-1.3), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9381-42)