IDO Monoclonal Antibody (eyedio), PerCP-eFluor™ 710, eBioscience™ - FAQs
IDO Monoclonal Antibody (eyedio), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IDO Monoclonal Antibody (eyedio), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9477-42, 46-9477-41)